Please login to your online account to display your discounted pricing

Mouse anti-IgM, Clone: SPM557, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442202

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes a protein of 75kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes.
Reconstitution Protein A purified
Clone SPM557
Concentration 1mg/mL
Antigen IgM
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 3507
Cross Reactivity Human
Immunogen Recombinant heavy chain of human IgM
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled