Please login to your online account to display your discounted pricing

Mouse anti-IgM, Clone: SPM557, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442201

  • / Each
 

Description & Specifications

Specifications

Format Purified
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM557
Isotype IgG1, κ
Concentration 1mg/mL
Purification Method Purified
Immunogen Recombinant heavy chain of human IgM
Regulatory Status RUO
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Test Specificity Recognizes a protein of 75kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes.
Primary or Secondary Primary
Cross Reactivity Human
Antigen IgM
Gene ID (Entrez) 3507
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Host Species Mouse