Please login to your online account to display your discounted pricing

Mouse anti-IgM, Clone: IM373, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen IgM
Clone IM373
Concentration 1mg/mL
Immunogen Recombinant heavy chain of human IgM
Gene Accession No. P01871
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23465001

 
novus biologicals canada
NBP23465001MG
0.1mg Each for $384.00

NBP23465002

 
novus biologicals canada
NBP23465002MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen IgM
Clone IM373
Concentration 1mg/mL
Immunogen Recombinant heavy chain of human IgM
Gene Accession No. P01871
Gene ID (Entrez) 3507
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting