Please login to your online account to display your discounted pricing

Mouse anti-IgM, Clone: IM373, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23465001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23465001

  • / Each
 

Description & Specifications

Specifications

Antigen IgM
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01871
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant heavy chain of human IgM
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3507
Test Specificity Recognizes a protein of 75kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Reconstitution Protein A purified
Concentration 1mg/mL
Cross Reactivity Human
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Clone IM373
Conjugate Unlabeled