Please login to your online account to display your discounted pricing

Mouse anti-IgM, Clone: IM260, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23425301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23425301

  • / Each
 

Description & Specifications

Specifications

Format Purified
Gene Accession No. P01871
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant heavy chain of human IgM
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3507
Cross Reactivity Human
Antigen IgM
Formulation PBS with 0.05% BSA.
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Protein A purified
Clone IM260
Concentration 0.2mg/mL
Conjugate Unlabeled