missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-IgM, Clone: IM260, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen IgM
Clone IM260
Concentration 1mg/mL
Immunogen Recombinant heavy chain of human IgM
Gene Accession No. P01871
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23464902 Novus Biologicals Canada
NBP23464902MG
0.2mg Each for $527.00
NBP23464901 Novus Biologicals Canada
NBP23464901MG
0.1mg Each for $384.00
Specifications Description & Specifications

Specifications

IgM
1mg/mL
P01871
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
IM260
Recombinant heavy chain of human IgM
3507
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot