Please login to your online account to display your discounted pricing

Mouse anti-IgM, Clone: IM260, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23464901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23464901

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Conjugate Unlabeled
Gene ID (Entrez) 3507
Format Purified
Cross Reactivity Human
Gene Accession No. P01871
Reconstitution Protein A purified
Immunogen Recombinant heavy chain of human IgM
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Antigen IgM
Primary or Secondary Primary
Isotype IgG1, κ
Clone IM260
Regulatory Status RUO
Concentration 1mg/mL
Quantity 0.1mg