Please login to your online account to display your discounted pricing

Mouse anti-IgM, Clone: DA4-4, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen IgM
Clone DA4-4
Concentration 0.2mg/mL
Immunogen Heavy chain of human IgM
Gene Accession No. P01871
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23425201

 
novus biologicals canada
NBP23425201ML
0.1mL Each for $139.00

NBP23425205

 
novus biologicals canada
NBP23425205ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen IgM
Clone DA4-4
Concentration 0.2mg/mL
Immunogen Heavy chain of human IgM
Gene Accession No. P01871
Gene ID (Entrez) 3507
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting