Please login to your online account to display your discounted pricing

Mouse anti-IgM, Clone: DA4-4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23425201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23425201

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Immunogen Heavy chain of human IgM
Primary or Secondary Primary
Antigen IgM
Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Reconstitution Protein A purified
Clone DA4-4
Concentration 0.2mg/mL
Cross Reactivity Human
Gene Accession No. P01871
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3507
Conjugate Unlabeled
Quantity 0.1mL