Please login to your online account to display your discounted pricing

Mouse anti-IgM, Clone: DA4-4, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23460101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23460101

  • / Each
 

Description & Specifications

Specifications

Immunogen Heavy chain of human IgM
Antigen IgM
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Quantity 0.1mg
Host Species Mouse
Regulatory Status RUO
Gene Accession No. P01871
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone DA4-4
Reconstitution Protein A purified
Purification Method Purified
Format Purified
Concentration 1mg/mL
Isotype IgG1, κ
Primary or Secondary Primary
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 3507