Please login to your online account to display your discounted pricing

Mouse anti-IgG2, Clone: 108G4.03, Novus Biologicals™

Manufacturer: novus biologicals canada  DDX5001B100005MG

 View all options for this product


For Research Use Only

Catalog No. DDX5001B05


Description & Specifications


Antigen IgG2
Applications Western Blotting
Clone 108G4.03
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation Tris-NaCl pH 8
Host Species Mouse
Immunogen Anti-Remicade(Infliximab)
Isotype IgG1
Purification Method Ion Exchange Chromatography
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3501