Please login to your online account to display your discounted pricing

Mouse anti-IgG2, Clone: 108G4.03, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX5001P100005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX5001P05

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Format Purified
Host Species Mouse
Immunogen Anti-Remicade(Infliximab)
Isotype IgG1
Regulatory Status RUO
Clone 108G4.03
Conjugate Unlabeled
Cross Reactivity Human
Antigen IgG2
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3501
Formulation Tris-NaCl pH 8
Purification Method Ion Exchange Chromatography
Quantity 0.05mg
Concentration 0.5mg/mL