Please login to your online account to display your discounted pricing

Mouse anti-IgG2, Clone: 107D8.04, Novus Biologicals™

$256.33 - $333.17

Specifications

Antigen IgG2
Clone 107D8.04
Concentration 0.5mg/mL
Immunogen Anti-Roactemra(Tocilizumab)
Gene ID (Entrez) 3501
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

DDX5020B05

 
novus biologicals canada
DDX5020B100005MG
0.05mg Each for $333.17

DDX5020P05

 
novus biologicals canada
DDX5020P100005MG
0.05mg Each for $256.33
Description & Specifications

Specifications

Antigen IgG2
Clone 107D8.04
Concentration 0.5mg/mL
Immunogen Anti-Roactemra(Tocilizumab)
Gene ID (Entrez) 3501
Formulation Tris-NaCl pH 8
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG2a
Conjugate Unlabeled
Format Purified
Purification Method Ion Exchange Chromatography
Applications Western Blot