Please login to your online account to display your discounted pricing

Mouse anti-IgG2, Clone: 107D8.04, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX5020P100005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX5020P05

  • / Each
 

Description & Specifications

Specifications

Antigen IgG2
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Cross Reactivity Human
Formulation Tris-NaCl pH 8
Host Species Mouse
Applications ELISA
Immunogen Anti-Roactemra(Tocilizumab)
Isotype IgG2a
Purification Method Ion Exchange Chromatography
Quantity 0.05mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3501
Clone 107D8.04
Concentration 0.5mg/mL
Conjugate Unlabeled