Please login to your online account to display your discounted pricing

Mouse anti-IgG2, Clone: 103A8.03, Novus Biologicals™

$309.05 - $332.66

Specifications

Antigen IgG2
Clone 103A8.03
Concentration 0.5mg/mL
Immunogen Anti-Remicade(Infliximab)
Gene ID (Entrez) 3501
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

DDX5002B05

 
novus biologicals canada
DDX5002B100005MG
0.05mg Each for $332.66

DDX5002P05

 
novus biologicals canada
DDX5002P100005MG
0.05mg Each for $309.05
Description & Specifications

Specifications

Antigen IgG2
Clone 103A8.03
Concentration 0.5mg/mL
Immunogen Anti-Remicade(Infliximab)
Gene ID (Entrez) 3501
Formulation Tris-NaCl pH 8
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Ion Exchange Chromatography
Applications ELISA