Please login to your online account to display your discounted pricing

Mouse anti-IgG2, Clone: 103A8.03, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX5002P100005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX5002P05

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications ELISA
Immunogen Anti-Remicade(Infliximab)
Purification Method Ion Exchange Chromatography
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Antigen IgG2
Formulation Tris-NaCl pH 8
Isotype IgG1
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3501
Clone 103A8.03
Conjugate Unlabeled
Concentration 0.5mg/mL