Please login to your online account to display your discounted pricing

Mouse anti-IgG2, Clone: 103A8.03, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX5002B100005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX5002B05

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Format Purified
Quantity 0.05mg
Formulation Tris-NaCl pH 8
Primary or Secondary Primary
Clone 103A8.03
Purification Method Ion Exchange Chromatography
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Immunogen Anti-Remicade(Infliximab)
Gene ID (Entrez) 3501
Monoclonal or Polyclonal Monoclonal
Antigen IgG2
Isotype IgG1
Concentration 0.5mg/mL