Please login to your online account to display your discounted pricing

Mouse anti-IgG, Clone: SPM566, Novus Biologicals™

$142.00 - $329.00

Specifications

Antigen IgG
Clone SPM566
Concentration 0.2mg/mL
Immunogen Purified human Ig Gamma Chain
Gene ID (Entrez) 3500
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23438101

 
novus biologicals canada
NBP23438101ML
0.1mL Each for $142.00

NBP23438105

 
novus biologicals canada
NBP23438105ML
0.5mL Each for $329.00
Description & Specifications

Specifications

Antigen IgG
Clone SPM566
Concentration 0.2mg/mL
Immunogen Purified human Ig Gamma Chain
Gene ID (Entrez) 3500
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting