Please login to your online account to display your discounted pricing

Mouse anti-IgG, Clone: SPM566, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438101

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 3500
Conjugate Unlabeled
Format Purified
Antigen IgG
Reconstitution Protein A purified
Cross Reactivity Human
Clone SPM566
Concentration 0.2mg/mL
Quantity 0.1mL
Isotype IgG2a, κ
Immunogen Purified human Ig Gamma Chain
Formulation PBS with 0.05% BSA.
Test Specificity Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes.
Purification Method Purified
Host Species Mouse