Please login to your online account to display your discounted pricing

Mouse anti-IgG, Clone: SPM566, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438105

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen IgG
Host Species Mouse
Immunogen Purified human Ig Gamma Chain
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.5mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3500
Test Specificity Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes.
Clone SPM566
Concentration 0.2mg/mL
Conjugate Unlabeled