Please login to your online account to display your discounted pricing

Mouse anti-IgG, Clone: SPM566, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen IgG
Clone SPM566
Concentration 1mg/mL
Immunogen Purified human Ig Gamma Chain
Gene ID (Entrez) 3500
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23442801

 
novus biologicals canada
NBP23442801MG
0.1mg Each for $384.00

NBP23442802

 
novus biologicals canada
NBP23442802MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen IgG
Clone SPM566
Concentration 1mg/mL
Immunogen Purified human Ig Gamma Chain
Gene ID (Entrez) 3500
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting