Please login to your online account to display your discounted pricing

Mouse anti-IgG, Clone: SPM566, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442802

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified human Ig Gamma Chain
Isotype IgG2a, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Antigen IgG
Quantity 0.2mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3500
Test Specificity Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes.
Reconstitution Protein A purified
Clone SPM566
Primary or Secondary Primary