Please login to your online account to display your discounted pricing

Mouse anti-IgG, Clone: SPM566, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442801

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified human Ig Gamma Chain
Isotype IgG2a, κ
Quantity 0.1mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Test Specificity Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes.
Clone SPM566
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Antigen IgG
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 3500
Reconstitution Protein A purified