Please login to your online account to display your discounted pricing

Mouse anti-IgG, Clone: IG266, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23425001ML

 View all options for this product


For Research Use Only

Catalog No. NBP23425001


Description & Specifications


Antigen IgG
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone IG266
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01857
Host Species Mouse
Immunogen Purified human Ig Gamma Chain
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3500
Reconstitution Protein A purified