Please login to your online account to display your discounted pricing

Mouse anti-IgG, Clone: ICO-97, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23425101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23425101

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3500
Cross Reactivity Human
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Conjugate Unlabeled
Antigen IgG
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01857
Host Species Mouse
Immunogen Purified human Ig Gamma Chain
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone ICO-97
Concentration 0.2mg/mL