Please login to your online account to display your discounted pricing

Mouse anti-IgG, Clone: ICO-97, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23425105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23425105

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Antigen IgG
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Immunogen Purified human Ig Gamma Chain
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3500
Concentration 0.2mg/mL
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01857
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone ICO-97
Conjugate Unlabeled