Please login to your online account to display your discounted pricing

Mouse anti-IgG, Clone: ICO-97, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23464701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23464701

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Conjugate Unlabeled
Host Species Mouse
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Quantity 0.1mg
Isotype IgG1, κ
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Reconstitution Protein A purified
Purification Method Purified
Gene Accession No. P01857
Clone ICO-97
Immunogen Purified human Ig Gamma Chain
Monoclonal or Polyclonal Monoclonal
Antigen IgG
Regulatory Status RUO
Gene ID (Entrez) 3500
Primary or Secondary Primary