Please login to your online account to display your discounted pricing

Mouse anti-IGF-I, Clone: SPM406, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23436101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23436101

  • / Each
 

Description & Specifications

Specifications

Purification Method Purified
Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Gene Accession No. P05019
Host Species Mouse
Regulatory Status RUO
Antigen IGF-I
Conjugate Unlabeled
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM406
Reconstitution Protein A purified
Quantity 0.1mL
Immunogen Purified human IGF-1 protein
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3479
Test Specificity This antibody is specific to Insulin-like Growth Factor (IGF-1) and shows minimal cross-reaction with IGF-11, Proinsulin, MSF, and Insulin.
Concentration 0.2mg/mL
Format Purified