Please login to your online account to display your discounted pricing

Mouse anti-IGF-I, Clone: SPM406, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440902

  • / Each
 

Description & Specifications

Specifications

Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blotting
Conjugate Unlabeled
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Primary or Secondary Primary
Quantity 0.2mg
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Gene ID (Entrez) 3479
Regulatory Status RUO
Format Purified
Reconstitution Protein A purified
Antigen IGF-I
Isotype IgG1, κ
Purification Method Purified
Clone SPM406
Gene Accession No. P05019
Test Specificity This antibody is specific to Insulin-like Growth Factor (IGF-1) and shows minimal cross-reaction with IGF-11, Proinsulin, MSF, and Insulin.
Immunogen Purified human IGF-1 protein