Please login to your online account to display your discounted pricing

Mouse anti-IGF-I, Clone: SPM406, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440901

  • / Each
 

Description & Specifications

Specifications

Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blotting
Conjugate Unlabeled
Antigen IGF-I
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P05019
Host Species Mouse
Immunogen Purified human IGF-1 protein
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3479
Test Specificity This antibody is specific to Insulin-like Growth Factor (IGF-1) and shows minimal cross-reaction with IGF-11, Proinsulin, MSF, and Insulin.
Clone SPM406
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG1, κ
Format Purified
Reconstitution Protein A purified
Concentration 1mg/mL