Please login to your online account to display your discounted pricing

Mouse anti-IGF-I, Clone: M23, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen IGF-I
Clone M23
Concentration 0.2mg/mL
Immunogen Purified human IGF-1 protein
Gene Accession No. P05019
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23424901

 
novus biologicals canada
NBP23424901ML
0.1mL Each for $139.00

NBP23424905

 
novus biologicals canada
NBP23424905ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen IGF-I
Clone M23
Concentration 0.2mg/mL
Immunogen Purified human IGF-1 protein
Gene Accession No. P05019
Gene ID (Entrez) 3479
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting