Please login to your online account to display your discounted pricing

Mouse anti-IGF-I, Clone: M23, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23424905ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23424905

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Antigen IGF-I
Formulation PBS with 0.05% BSA.
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 3479
Concentration 0.2mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Gene Accession No. P05019
Immunogen Purified human IGF-1 protein
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone M23
Conjugate Unlabeled