Please login to your online account to display your discounted pricing

Mouse anti-IGF-I, Clone: M23, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23424901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23424901

  • / Each
 

Description & Specifications

Specifications

Antigen IGF-I
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P05019
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified human IGF-1 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3479
Reconstitution Protein A purified
Clone M23
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Regulatory Status RUO