missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-IGF-I, Clone: M23, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen IGF-I
Clone M23
Concentration 1mg/mL
Immunogen Purified human IGF-1 protein
Gene Accession No. P05019
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23467901 Novus Biologicals Canada
NBP23467901MG
0.1mg Each for $384.00
NBP23467902 Novus Biologicals Canada
NBP23467902MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

IGF-I
1mg/mL
P05019
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
M23
Purified human IGF-1 protein
3479
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot