Please login to your online account to display your discounted pricing

Mouse anti-IGF-I, Clone: M23, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467902

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Antigen IGF-I
Immunogen Purified human IGF-1 protein
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Gene ID (Entrez) 3479
Format Purified
Host Species Mouse
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Primary or Secondary Primary
Isotype IgG1, κ
Purification Method Purified
Concentration 1mg/mL
Regulatory Status RUO
Quantity 0.2mg
Reconstitution Protein A purified
Clone M23
Gene Accession No. P05019
Monoclonal or Polyclonal Monoclonal