Please login to your online account to display your discounted pricing

Mouse anti-IGF-I, Clone: M23, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467901

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Quantity 0.1mg
Host Species Mouse
Gene ID (Entrez) 3479
Purification Method Purified
Gene Accession No. P05019
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Monoclonal or Polyclonal Monoclonal
Isotype IgG1, κ
Concentration 1mg/mL
Antigen IGF-I
Clone M23
Primary or Secondary Primary
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Immunogen Purified human IGF-1 protein
Conjugate Unlabeled