Please login to your online account to display your discounted pricing

Mouse anti-ICOS, Clone: ISA-3, FITC, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen ICOS
Clone ISA-3
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was ICOS.
Gene ID (Entrez) 29851
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143917

 
novus biologicals canada
NBP143917100TESTS
100 Tests Each for $329.00

NBP14391725

 
novus biologicals canada
NBP14391725TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen ICOS
Clone ISA-3
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was ICOS.
Gene ID (Entrez) 29851
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry