Please login to your online account to display your discounted pricing

Mouse anti-ICOS, Clone: ISA-3, Biotin, Novus Biologicals™

$175.00 - $395.00

Specifications

Antigen ICOS
Clone ISA-3
Immunogen The immunogen for this antibody was ICOS.
Gene ID (Entrez) 29851
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14347201

 
novus biologicals canada
NBP14347201MG
0.1mg Each for $395.00

NBP14347203

 
novus biologicals canada
NBP1434720025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen ICOS
Clone ISA-3
Immunogen The immunogen for this antibody was ICOS.
Gene ID (Entrez) 29851
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry