Please login to your online account to display your discounted pricing

Mouse anti-ICAM-3/CD50, Clone: SPM505, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen ICAM-3/CD50
Clone SPM505
Concentration 0.2mg/mL
Immunogen Stimulated human leukocytes
Gene Accession No. P32942
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23282601

 
novus biologicals canada
NBP23282601ML
0.1mL Each for $139.00

NBP23282610

 
novus biologicals canada
NBP23282610ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen ICAM-3/CD50
Clone SPM505
Concentration 0.2mg/mL
Immunogen Stimulated human leukocytes
Gene Accession No. P32942
Gene ID (Entrez) 3385
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting