Please login to your online account to display your discounted pricing

Mouse anti-ICAM-3/CD50, Clone: SPM505, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23282610ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23282610

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Quantity 1.0mL
Isotype IgG2b, κ
Monoclonal or Polyclonal Monoclonal
Clone SPM505
Antigen ICAM-3/CD50
Format Purified
Formulation PBS with 0.05% BSA.
Conjugate Unlabeled
Host Species Mouse
Immunogen Stimulated human leukocytes
Gene Accession No. P32942
Gene ID (Entrez) 3385
Reconstitution Protein A purified
Concentration 0.2mg/mL
Cross Reactivity Human