Please login to your online account to display your discounted pricing

Mouse anti-ICAM-3/CD50, Clone: SPM505, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23282601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23282601

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Stimulated human leukocytes
Antigen ICAM-3/CD50
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P32942
Host Species Mouse
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3385
Reconstitution Protein A purified
Clone SPM505
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human