Please login to your online account to display your discounted pricing

Mouse anti-ICAM-3/CD50, Clone: SPM505, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23477602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23477602

  • / Each
 

Description & Specifications

Specifications

Quantity 0.2mg
Gene ID (Entrez) 3385
Clone SPM505
Cross Reactivity Human
Purification Method Purified
Test Specificity Recognizes the D1 domain of an N-glycosylated glycoprotein of 120kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Stimulated human leukocytes
Antigen ICAM-3/CD50
Format Purified
Gene Accession No. P32942
Primary or Secondary Primary
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Reconstitution Protein A purified
Isotype IgG2b, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Formulation 10mM PBS. Does not contain BSA. Azide Free.