Please login to your online account to display your discounted pricing

Mouse anti-ICAM-3/CD50, Clone: SPM505, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23477601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23477601

  • / Each
 

Description & Specifications

Specifications

Antigen ICAM-3/CD50
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Stimulated human leukocytes
Purification Method Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Test Specificity Recognizes the D1 domain of an N-glycosylated glycoprotein of 120kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3.
Clone SPM505
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG2b, κ
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Concentration 1mg/mL
Cross Reactivity Human
Gene Accession No. P32942
Host Species Mouse
Primary or Secondary Primary
Gene ID (Entrez) 3385