Please login to your online account to display your discounted pricing

Mouse anti-ICAM-3/CD50, Clone: 186-2G9, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23452102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23452102

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Isotype IgG2b, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.2mg
Regulatory Status RUO
Clone 186-2G9
Format Purified
Gene Accession No. P32942
Monoclonal or Polyclonal Monoclonal
Antigen ICAM-3/CD50
Reconstitution Protein A purified
Host Species Mouse
Concentration 1mg/mL
Immunogen Stimulated human leukocytes
Cross Reactivity Human
Gene ID (Entrez) 3385
Conjugate Unlabeled
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.