Please login to your online account to display your discounted pricing

Mouse anti-ICAM-3/CD50, Clone: 186-2G9, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23452101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23452101

  • / Each
 

Description & Specifications

Specifications

Antigen ICAM-3/CD50
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone 186-2G9
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P32942
Host Species Murine
Immunogen Stimulated human leukocytes
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3385
Reconstitution Protein A purified
Target Species Human