Please login to your online account to display your discounted pricing

Mouse anti-ICAM-1/CD54, Clone: HA58, PE, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen ICAM-1/CD54
Clone HA58
Immunogen The immunogen for this antibody was CD54.
Gene ID (Entrez) 3383
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200243

 
novus biologicals canada
NBP200243100TESTS
100 Tests Each for $296.00

NBP20024325

 
novus biologicals canada
NBP20024325TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen ICAM-1/CD54
Clone HA58
Immunogen The immunogen for this antibody was CD54.
Gene ID (Entrez) 3383
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry