Please login to your online account to display your discounted pricing

Mouse anti-ICAM-1/CD54, Clone: HA58, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen ICAM-1/CD54
Clone HA58
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD54.
Gene ID (Entrez) 3383
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14339701

 
novus biologicals canada
NBP14339701MG
0.1mg Each for $285.00

NBP14339703

 
novus biologicals canada
NBP1433970025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen ICAM-1/CD54
Clone HA58
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD54.
Gene ID (Entrez) 3383
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting