Please login to your online account to display your discounted pricing

Mouse anti-ICAM-1/CD54, Clone: HA58, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14339701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14339701

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Host Species Mouse
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen The immunogen for this antibody was CD54.
Isotype IgG1, κ
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Clone HA58
Concentration 0.5mg/mL
Conjugate Unlabeled
Antigen ICAM-1/CD54
Formulation Phosphate buffer pH 7.2
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3383
Cross Reactivity Human