Please login to your online account to display your discounted pricing

Mouse anti-ICAM-1/CD54, Clone: HA58, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433970025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14339703

  • / Each
 

Description & Specifications

Specifications

Antigen ICAM-1/CD54
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Isotype IgG1, κ
Format Affinity Purified
Quantity 0.025mg
Host Species Mouse
Formulation Phosphate buffer pH 7.2
Gene ID (Entrez) 3383
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Clone HA58
Immunogen The immunogen for this antibody was CD54.