Please login to your online account to display your discounted pricing

Mouse anti-ICAM-1/CD54, Clone: HA58, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen ICAM-1/CD54
Clone HA58
Immunogen The immunogen for this antibody was CD54.
Gene ID (Entrez) 3383
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14363001

 
novus biologicals canada
NBP14363001MG
0.1mg Each for $252.00

NBP14363003

 
novus biologicals canada
NBP1436300025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen ICAM-1/CD54
Clone HA58
Immunogen The immunogen for this antibody was CD54.
Gene ID (Entrez) 3383
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry